Learn More
PCDHAC1 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24314-1-AP-150UL
 View more versions of this product
Product Code. 18024735
  • € 410.00 / 150µL
Description
The PCDHAC1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PCDHAC1. This antibody recognizes human, mouse antigen. The PCDHAC1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
PCDHAC1
250 μg/mL
Rabbit
Primary
Human, Mouse
150 kDa
IgG
Western Blot, ELISA
PCDH ALPHA C1, PCDHAC1, Protocadherin alpha C1
150 μL
56135
Store at -20°C. Stable for one year after shipment.