missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PCDHB13 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15808013
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15808013 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15808013 Supplier Invitrogen™ Supplier No. PA569599

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Human: 100%.

Potential calcium-dependent cell-adhesion protein. May be involved in the establishment and maintenance of specific neuronal connections in the brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDHB13
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PCDHB13
Gene Accession No. Q9Y5F0
Gene Alias Pcdbh6; PCDHB13; PCDH-BETA13; PCDH-beta-13; PcdhbM; protocadherin beta 13; protocadherin beta-13; protocadherin beta-6; UNQ332/PRO531
Gene Symbols PCDHB13
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human PCDHB13.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56123
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.