missing translation for 'onlineSavingsMsg'
Learn More

PCDHB3, Mouse anti-Human, Clone: 4F6, Abnova™

Product Code. 16158656
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16158656 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16158656 Supplier Abnova Supplier No. H00056132M01.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sequence: ASEEIRKTFQLNPITGDMQLVKYLNFEAINSYEVDIEAKDGGGLSGKSTVIVQVVDVNDNPPELTLSSVNSPIPENSGETVLAVFSVSDLDSGDNGRV

Specifications

Antigen PCDHB3
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4F6
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene protocadherin beta 3
Gene Accession No. NM_018937
Gene Alias PCDH-BETA3
Gene Symbols PCDHB3
Host Species Mouse
Immunogen PCDHB3 (NP_061760, 284 a.a. ∼ 381 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56132
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.