missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCDHB8 Monoclonal Antibody (OTI1E1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812705
This item is not returnable.
View return policy
Description
Calcium-dependent cell-adhesion protein involved in cells self-recognition and non-self discrimination. Thereby, it is involved in the establishment and maintece of specific neuronal connections in the brain.Specifications
| PCDHB8 | |
| Monoclonal | |
| Unconjugated | |
| PCDHB8 | |
| PCDH-BETA8, PCDH3I | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 56128 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI1E1 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UN66 | |
| PCDHB8 | |
| Human recombit protein fragment corresponding to amino acids 29-418 of human Pcdhb8 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction