missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCDHGA1 Antibody (S159-5), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42207G
This item is not returnable.
View return policy
Description
PCDHGA1 Monoclonal specifically detects PCDHGA1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| PCDHGA1 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PCDHGA1 | |
| Fusion protein corresponding to amino acids 808-931 (C-terminal cytoplasmic constant domain) of mouse Protocadherin-gamma-A1 that is shared by all 22 Gamma-protocadherins (A subfamily amino acids ∼807-930, B subfamily amino acids ∼789-912 and C subfamily amino acids ∼818-941). 99% identity with human (123/124 amino acids). | |
| 0.1 mL | |
| 56114 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| S159-5 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| MGC138287, MGC138289, PCDH-GAMMA-A1, protocadherin gamma subfamily A, 1, protocadherin gamma-A1 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Detects approx 100kDa. Cross-reacts with all Gamma-protocadherin-A, -B and -C proteins. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction