missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pcdhga1 Mouse anti-Human, Mouse, Rat, Biotin, Clone:S159-5, Abnova™
Mouse monoclonal antibody raised against partial recombinant mouse Pcdhga1.
Brand: Abnova MAB16865.100ug
This item is not returnable.
View return policy
Specifications
| Pcdhga1 | |
| Monoclonal | |
| Biotin | |
| Immunocytochemistry (1:100) Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Western Blot (1:1000) The optimal working dilution should be determined by the end user. |
|
| Pcdhga1 | |
| Recombinant protein corresponding to amino acids 808-931 at C-terminus of mouse Pcdhga1. | |
| 93709 | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| S159-5 | |
| protocadherin gamma subfamily A, 1 | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
| Mouse | |
| 100 μg | |
| This antibody crossreacts with all Gamma-protocadherin-A, -B and -C proteins. | |
| Store at -20°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction