missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCDHGA10, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00056106-B01.50uL
This item is not returnable.
View return policy
Description
Sequence: MAAQRNRSKESKDCSGLVLLCLFFGIPWEAGARQISYSIPEELEKGSFVGNISKDLGLAPRELAERGVRIVSRGRTQLFSLNPRSGSLITAGRIDREELCAQSARCVVSFNILVEDRVKLFGIEIEVTDINDNAPKFQAENLDVKINENVAAGMRFPLPEAIDPDVGVNSLQSYQLSPNKHFSLRVQSRANGVKYPELVLEHSLDREEEAIHHLVLTASDGGDPLRSGTVLVSVTVFDANDNAPVFTLPEYRVSVPENLPVGTQLLTVTATDRDEGANGEVTYSFRKLPDTQLLKFQLNKYTGEIKISENLDYEETGFYEIEIQAEDGGAYLATAKVLITVEDVNDNSPELTITSLFSPVTEDSPLGTVVALLNVHDLDSEQNGQVTCSILAYLPFKLEKSIDSYYRLVIHRALDREQVSSYNITVTATDGGSPPLSTEAHFMLQVADINDNPPTFSQVSYFTYIPENNARGASIFSVTALDPDSKENAQIIYSLAEDTIQGVPLSSYISINSDTGVLYALRSFDYEQFHELQMQVTASDSGDPPLSSNVSLSLFVLDQNDNAPEILYPALPTDGSTGVELAPRSAEPGYLVTKVVAVDRDSGQNAWLSYRLLKASEPGLFAVGEHTGEVRTARALLDRDALKQSLVVAVQDHGQPPLSATVTLTVAVADSIPQVLADLGSFESPANSETSDLTLYLVVAVAAVSCVFLAFVIVLLAHRLRRWHKSRLLQASGGGLTGVSGSHFVGVDGVRAFLQTYSHEVSLTADSRKSHLIFPQPNYADTLISQESCEKNDPLSLLDDSKFPIEDTPLVPVSFIFIFTFVKKKKIGFYFEVCGMMVESVNAKTLMSRISpecifications
| PCDHGA10 | |
| Polyclonal | |
| No additive | |
| NM_032090.1 | |
| PCDHGA10 | |
| PCDHGA10 (NP_114479.1, 1 a.a. ∼ 850 a.a) full-length human protein. | |
| RUO | |
| 56106 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| protocadherin gamma subfamily A, 10 | |
| PCDH-GAMMA-A10 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction