missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCDHGC5 Antibody (OTI2F3), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73270APC
This item is not returnable.
View return policy
Description
PCDHGC5 Monoclonal antibody specifically detects PCDHGC5 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PCDHGC5 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 56097 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot | |
| OTI2F3 | |
| Western Blot | |
| MGC138286, MGC138288, PCDH-GAMMA-C5, protocadherin gamma subfamily C, 5, protocadherin gamma-C5 | |
| Human recombinant protein fragment corresponding to amino acids 35-258 of human PCDHGC5(NP_115783) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction