missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCDHGC5 Monoclonal Antibody (OTI1A6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807813S
This item is not returnable.
View return policy
Description
PCDHGC5 Monoclonal antibody specifically detects PCDHGC5 in Human samples. It is validated for Western BlotSpecifications
| PCDHGC5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y5F6 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 56097 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1A6 | |
| Unconjugated | |
| PCDHGC5 | |
| PCDH-gamma-C5; PCDHGC5; protocadherin gamma c5; protocadherin gamma subfamily C, 5; protocadherin gamma-C5 | |
| Human recombinant protein fragment corresponding to amino acids 35-258 of human PCDHGC5 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction