missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCF11 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-12787AF532
This item is not returnable.
View return policy
Description
PCF11 Polyclonal antibody specifically detects PCF11 in Human samples. It is validated for Western Blot
Specifications
| PCF11 | |
| Polyclonal | |
| Western Blot | |
| KIAA0824PCF11p homolog, PCF11, cleavage and polyadenylation factor II subunit, homolog, PCF11, cleavage and polyadenylation factor II subunit, homolog (S. cerevisiae), PCF11, cleavage and polyadenylation factor subunit, homolog (S. cerevisiae), pre-mRNA cleavage complex 2 protein Pcf11, Pre-mRNA cleavage complex II protein Pcf11 | |
| The immunogen this antibody was made to, maps to a region between residue 1505 and 1555 of human Cleavage and Polyadenylation Factor II Subunit using the numbering given in entry NP_056969.2 (GeneID 51585). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 51585 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction