missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCGF3 Antibody (PCRP-PCGF3-1D5), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24070DL594
This item is not returnable.
View return policy
Description
PCGF3 Monoclonal antibody specifically detects PCGF3 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| PCGF3 | |
| Monoclonal | |
| DyLight 594 | |
| DKFZp686D20235, DONG1, FLJ36550, FLJ43813, MGC129615, MGC40413, polycomb group ring finger 3, polycomb group RING finger protein 3, ring finger protein 3, RNF3, RNF3ARING finger protein 3A | |
| Recombinant full-length human PCGF3 protein | |
| 0.1 mL | |
| Zinc Finger | |
| 10336 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| PCRP-PCGF3-1D5 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction