missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCGF5 Antibody (3C10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00084333-M01
This item is not returnable.
View return policy
Description
PCGF5 Monoclonal antibody specifically detects PCGF5 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA
Specifications
| PCGF5 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| MGC16202, polycomb group ring finger 5, polycomb group RING finger protein 5, ring finger protein (C3HC4 type) 159, RING finger protein 159, RNF159 | |
| PCGF5 (NP_115749.2, 51 a.a. ∽ 148 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. NDCPRCGNQVHETNPLEMLRLDNTLEEIIFKLVPGLREQELERESEFWKKNKPQENGQDDTSKADKPKVDEEGDENEDDKDYHRSDPQIAICLDCLRN | |
| 0.1 mg | |
| Zinc Finger | |
| 84333 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Sandwich ELISA | |
| 3C10 | |
| Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA | |
| NP_115749 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction