missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCGF5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558353
This item is not returnable.
View return policy
Description
Immunogen sequence: TCIVQHFEDS NDCPRCGNQV HETNPLEMLR LDNTLEEIIF KLVPGLREQE LERESEFWKK NKPQENGQDD TSKAD Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 93%.
Component of a Polycomb group (PcG) multiprotein PRC1-like complex, a complex class required to maintain the transcriptionally repressive state of many genes, including Hox genes, throughout development. PcG PRC1 complex acts via chromatin remodeling and modification of histones; it mediates monoubiquitination of histone H2A 'Lys-119', rendering chromatin heritably changed in its expressibility (PubMed:26151332). Within the PRC1-like complex, regulates RNF2 ubiquitin ligase activity (PubMed:26151332). Plays a redundant role with PCGF3 as part of a PRC1-like complex that mediates monoubiquitination of histone H2A 'Lys-119' on the X chromosome and is required for normal silencing of one copy of the X chromosome in XX females. [UniProt].
Specifications
| PCGF5 | |
| Polyclonal | |
| Unconjugated | |
| PCGF5 | |
| 0610009F02Rik; 1110054A01Rik; 5830406C17Rik; 5830443C21Rik; 9530023M17Rik; AI324127; Pcgf5; polycomb group ring finger 5; polycomb group RING finger protein 5; polycomb group RING finger protein 5-like protein; ring finger protein (C3HC4 type) 159; RING finger protein 159; Rnf159 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84333 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86SE9 | |
| PCGF5 | |
| Recombinant protein corresponding to Human PCGF5. Recombinant protein control fragment (Product #RP-97187). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction