Learn More
PCI-34051, MedChemExpress
Product Code. 16442487
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16442487
Brand: Medchem Express HY15224
MedChemExpress PCI-34051 is a potent and selective HDAC8 inhibitor with IC50 of 10 nM, with >200-fold selectivity over the other HDAC isoforms.
PCI-34051 is a potent and selective HDAC8 inhibitor with IC50 of 10 nM, with >200-fold selectivity over the other HDAC isoforms.
Specifications
Chemical Name or Material PCI-34051
CAS 950762-95-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H16N2O3
Quantity 50 mg
Solubility Information DMSO : ≥ 30 mg/mL (101.24 mM)
SMILES COC(C=C1)=CC=C1CN2C3=CC(C(NO)=O)=CC=C3C=C2
Molecular Weight (g/mol) 296.32
Formula Weight 296.32
Percent Purity 97.28%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only