missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCIF1 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97683JF549
This item is not returnable.
View return policy
Description
PCIF1 Polyclonal antibody specifically detects PCIF1 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| PCIF1 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| bA465L10.1, C20orf67, chromosome 20 open reading frame 67, PDX-1 C terminus-interacting factor 1, PDX1 C-terminal inhibiting factor 1, phosphorylated CTD-interacting factor 1 | |
| Produced in rabbits immunized with E. coli-derived Human PCIF1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 63935 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction