missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pclo Mouse anti-Human, Mouse, Rat, FITC, Clone:6H9-B6, Abnova™
Mouse monoclonal antibody raised against full length recombinant mouse Pclo.
Brand: Abnova MAB16779.100ug
This item is not returnable.
View return policy
Specifications
| Pclo | |
| Monoclonal | |
| FITC | |
| ELISA Immunocytochemistry (1:100) Immunofluorescence (1:100) Western Blot (1:1000) The optimal working dilution should be determined by the end user. |
|
| Pclo | |
| Mouse | |
| 100 μg | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a κ |
| ELISA, Immunocytochemistry, Immunofluorescence, Western Blot | |
| 6H9-B6 | |
| piccolo (presynaptic cytomatrix protein) | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
| Acz, Pico, mKIAA0559 | |
| Recombinant protein corresponding to full length mouse Pclo. | |
| 26875 | |
| Store at -20°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction