Learn More
PCM1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 19856-1-AP-150UL
 View more versions of this product
Product Code. 16889334
  • € 410.00 / 150µL
Description
The protein encoded by this gene is a component of centriolar satellites, which are electron dense granules scattered around centrosomes. Inhibition studies show that this protein is essential for the correct localization of several centrosomal proteins, and for anchoring microtubules to the centrosome. Chromosomal aberrations involving this gene are associated with papillary thyroid carcinomas and a variety of hematological maligcies, including atypical chronic myeloid leukemia and T-cell lymphoma.
Specifications
Specifications
PCM1
Polyclonal
Unconjugated
PCM1
hPCM 1, PCM 1, PCM1, pericentriolar material 1, PTC4
Rabbit
Antigen Affinity Chromatography
RUO
18536, 5108
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q15154, Q9R0L6
PCM1
PCM1 Fusion Protein Ag13929
150 μL
Primary
Human, Mouse