missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCMT1 Monoclonal Antibody (OTI2E7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809919
This item is not returnable.
View return policy
Description
Three classes of protein carboxyl methyltransferases, distinguished by their methyl-acceptor substrate specificity, have been found in prokaryotic and eukaryotic cells. The type II enzyme catalyzes the transfer of a methyl group from S-adenosyl-L-methionine to the free carboxyl groups of D-aspartyl and L-isoaspartyl residues. These methyl-accepting residues result from the spontaneous deamidation, isomerization, and racemization of normal L-aspartyl and L-asparaginyl residues and represent sites of covalent damage to aging proteins PCMT1 (EC 2.1.1.77) is a protein repair enzyme that initiates the conversion of abnormal D-aspartyl and L-isoaspartyl residues to the normal L-aspartyl form.Specifications
| PCMT1 | |
| Monoclonal | |
| Unconjugated | |
| PCMT1 | |
| PIMT | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5110 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI2E7 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P22061 | |
| PCMT1 | |
| Human recombit protein fragment corresponding to amino acids 60-285 of human PCMT1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction