missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCNA Antibody (5E6/2), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50238AF594
This item is not returnable.
View return policy
Description
PCNA Monoclonal specifically detects PCNA in Yeast samples. It is validated for Western Blot, ELISA.
Specifications
| PCNA | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| PCNA | |
| Purified recombinant His6-PCNA from budding yeast, Saccharomyces cerevisiae, produced in Escherichia coli | |
| 0.1 mL | |
| 5111 | |
| Store at 4°C in the dark. |
| Western Blot, ELISA | |
| 5E6/2 | |
| Western Blot, ELISA | |
| cyclin, DNA polymerase delta auxiliary protein, MGC8367, proliferating cell nuclear antigen | |
| Mouse | |
| Protein A purified | |
| Primary | |
| Yeast | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction