missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCNA Antibody (PCNA/694) - IHC-Prediluted, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-48284
This item is not returnable.
View return policy
Description
PCNA Monoclonal specifically detects PCNA in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| PCNA | |
| Monoclonal | |
| Unconjugated | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| PCNA | |
| Recombinant full length human PCNA protein (Uniprot: P12004) | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Recognizes a non-histone protein of 36kDa, which is identified as proliferating cell nuclear antigen (PCNA). It is also known as cyclin or polymerase delta auxiliary protein. Elevated expression of PCNA/cyclin has been shown in the nucleus during late G1 phase immediately before the onset of DNA synthesis, becoming maximal during S-phase and declining during G2 and M phases. This monoclonal antibody is excellent for multiple applications. | |
| Store at 4C. | |
| IgG2a κ |
| Immunohistochemistry (Paraffin) | |
| PCNA/694 | |
| Immunohistochemistry-Paraffin | |
| cyclin, DNA polymerase delta auxiliary protein, MGC8367, proliferating cell nuclear antigen | |
| Mouse | |
| 36 kDa | |
| 7 mL | |
| Autophagy, Base Excision Repair, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Polymerases, DNA Repair, Loading Controls, Stem Cell Markers | |
| 5111 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction