Learn More
PCNA Monoclonal Antibody (OTI6A10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA800975S
Encompass_Preferred
 View more versions of this product
Product Code. 17549202
  • € 242.00 / 30µL
Description
PCNA Monoclonal antibody specifically detects PCNA in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
PCNA
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P04818
Mouse
Affinity chromatography
RUO
7298
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI6A10
Unconjugated
TYMS
ATLD2; cb16; Cyclin; DNA polymerase delta auxiliary protein; etID36690.10; fa28e03; fb36g03; HGCN8729; hypothetical protein LOC515499; MGC8367; Pcna; pcna protein; Pcna/cyclin; PCNAR; POL30; Proliferating cell nuclear antigen; wu:fa28e03; wu:fb36g03; YBR0811; YBR088C
Full length human recombinant protein of human TYMS produced in SF9 cell.