missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCNA Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI2G7, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800875
This item is not returnable.
View return policy
Description
PCNA (polymerase delta auxiliary protein) is essential for DNA replication and is involved in DNA excision and mismatch repair pathways. PCNA binds to the CDK inhibitor p21, the structure-specific endonucleases Fen1 and XPG, and DNA cytosine 5-methyltransferase (MCMT). PCNA is a potentially useful marker of cells with proliferative potential and for identifying the proliferation status of tumor tissue (i.e. relevant to prognosis). PCNA is a marker for cells in early G1 phase and S phase of the cell cycle. PCNA is found in the nucleus and is a cofactor of DNA polymerase delta, and acts as a homotrimer and helps increase the processing of leading strand synthesis during DNA replication. In response to DNA damage, PCNA is ubiquitinated and is involved in the RAD6-dependent DNA repair pathway. Two transcript variants encoding the same protein have been found for PCNA. Pseudogenes of PCNA have been described on chromosome 4 and on the X chromosome.Specifications
| PCNA | |
| Monoclonal | |
| Unconjugated | |
| PCNA | |
| ATLD2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 18538, 25737, 477166, 5111 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI2G7 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P04961, P12004, P17918 | |
| Pcna | |
| Full length human recombit protein of human PCNA produced in E.coli. | |
| 100 μL | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction