missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCNA Mouse, Clone: SPM350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-32989-0.02MG
This item is not returnable.
View return policy
Description
PCNA Monoclonal specifically detects PCNA in Human, Mouse, Rat, Porcine, Chicken, Drosophila, Monkey, S. pombe, Yeast, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| PCNA | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.05% BSA. with 0.05% Sodium Azide | |
| cyclin, DNA polymerase delta auxiliary protein, MGC8367, proliferating cell nuclear antigen | |
| Mouse | |
| 36 kDa | |
| 0.02 mg | |
| Autophagy, Base Excision Repair, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Polymerases, DNA Repair, Phospho Specific, Stem Cell Markers | |
| 5111 | |
| Human, Mouse, Rat, Pig, Chicken, Drosophila, Primate, Yeast, Zebrafish | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| SPM350 | |
| Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1.0ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml | |
| P12004 | |
| PCNA | |
| Rat PCNA/Protein A fusion protein | |
| Protein A purified | |
| RUO | |
| Primary | |
| Recognizes a non-histone protein of 36kDa, which is identified as proliferating cell nuclear antigen (PCNA). It is also known as cyclin or polymerase delta auxiliary protein. Elevated expression of PCNA/cyclin has been shown in the nucleus during late G1 phase immediately before the onset of DNA synthesis, becoming maximal during S-phase and declining during G2 and M phases. This MAb is excellent for multiple applications. | |
| Store at 4C. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction