Learn More
PCNP Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-37692H
Code produit. 18666705
  • € 633.00 / 0.10mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
PCNP Polyclonal antibody specifically detects PCNP in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Spécification
Spécification
PCNP
Polyclonal
Western Blot
DKFZp781I24156, PEST proteolytic signal containing nuclear protein, PEST proteolytic signal-containing nuclear protein, PEST-containing nuclear protein
This antibody maps to a region between residue 1 to 50 of human PEST Proteolytic Signal Containing Nuclear Protein using the numbering given in entry NP_065090.1 (GeneID 57092).
0.1 mL
Cell Cycle and Replication
57092
Store at 4°C in the dark.
IgG
Western Blot
HRP
PBS
Rabbit
Immunogen affinity purified