missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCNP Antibody (OTI4E3) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73272
This item is not returnable.
View return policy
Description
PCNP Monoclonal specifically detects PCNP in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| PCNP | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:2000 | |
| DKFZp781I24156, PEST proteolytic signal containing nuclear protein, PEST proteolytic signal-containing nuclear protein, PEST-containing nuclear protein | |
| Mouse | |
| 18.7 kDa | |
| 100 μg | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot | |
| OTI4E3 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| PCNP | |
| Full length human recombinant protein of human PCNP(NP_065090) produced in E.coli. | |
| Affinity Purified | |
| Cell Cycle and Replication | |
| 57092 | |
| Human, Mouse, Rat | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction