missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCOLCE Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583637
This item is not returnable.
View return policy
Description
Immunogen sequence: PKSQPPEKTE ESPSAPDAPT CPKQCRRTGT LQSNFCASSL VVTATVKSMV REPGEGLAVT VSLIGAYKTG GLDLPS Highest antigen sequence indentity to the following orthologs: Mouse - 71%, Rat - 71%.
Fibrillar collagen types I-III are synthesized as precursor molecules known as procollagens. These precursors contain amino- and carboxyl-terminal peptide extensions known as N- and C-propeptides, respectively, which are cleaved, upon secretion of procollagen from the cell, to yield the mature triple helical, highly structured fibrils. This gene encodes a glycoprotein which binds and drives the enzymatic cleavage of type I procollagen and heightens C-proteinase activity.
Specifications
| PCOLCE | |
| Polyclonal | |
| Unconjugated | |
| PCOLCE | |
| astroglial cell transcript 2; Astt2; Astt-2; P14; Pcolce; PCPE; PCPE1; PCPE-1; procollagen C-endopeptidase enhancer; procollagen C-endopeptidase enhancer 1; procollagen C-endopeptidase enhancer protein; procollagen COOH-terminal proteinase enhancer 1; procollagen C-proteinase enhancer 1; procollagen C-proteinase enhancer protein; procollagen, type 1, COOH-terminal proteinase enhancer; type 1 procollagen C-proteinase enhancer protein; Type I procollagen COOH-terminal proteinase enhancer | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5118 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15113 | |
| PCOLCE | |
| Recombinant protein corresponding to Human PCOLCE. Recombinant protein control fragment (Product #RP-104765). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction