Learn More
PCP4 Antibody (CL5306) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61410C
Product Code. 30291508
  • € 737.00 / 0.10mL
Description
PCP4 Monoclonal antibody specifically detects PCP4 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PCP4
Monoclonal
DyLight 650
brain specific polypeptide PEP19, 10Brain-specific antigen PCP-4, Brain-specific polypeptide PEP-19, PEP19, PEP-19, Purkinje cell protein 4
This antibody was developed using a recombinant protein derived from P48539, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human, Mouse, Rat
Purified
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL5306
50mM Sodium Borate
Mouse
Protein A purified
RUO
5121
Store at 4⪚C in the dark.