missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCPTP1 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 213.00 - € 507.00
Specifications
| Antigen | PCPTP1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30230004
|
Novus Biologicals
NBP3-33262-20ul |
20 μL |
€ 213.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30229965
|
Novus Biologicals
NBP3-33262-100ul |
100 μL |
€ 507.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PCPTP1 Monoclonal antibody specifically detects PCPTP1 in Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| PCPTP1 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 5801 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| Ch-1 PTPase, ch-1PTPase, DKFZp781C1038, EC 3.1.3.48, ECPTP, EC-PTP, FLJ34328, MGC131968, MGC148170, NC-PTPCOM1, PCPTP1, protein tyrosine phosphatase Cr1PTPase, protein tyrosine phosphatase, receptor type, R, protein-tyrosine phosphatase NC-PTPCOM1, Protein-tyrosine phosphatase PCPTP1, PTPBR7, PTPRQ, PTP-SL, receptor-type tyrosine-protein phosphatase R, R-PTP-R | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PCPTP1 (Q15256).,, Sequence:, MRRAVCFPALCLLLNLHAAGCFSGNNDHFLAINQKKSGKPVFIYKHSQDIEKSLDIAPQKIYRHSYHSSSEAQVSKRHQIVNSAFPRPAYDPSLNLLAMD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title