missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PCSK1N Polyclonal Antibody
GREENER_CHOICE

Product Code. 15848833
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15848833 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15848833 Supplier Invitrogen™ Supplier No. PA570424

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rat: 100%.

The protein encoded by this gene functions as an inhibitor of prohormone convertase 1, which regulates the proteolytic cleavage of neuroendocrine peptide precursors. The proprotein is further processed into multiple short peptides. A polymorphism within this gene may be associated with obesity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCSK1N
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PCSK1N
Gene Accession No. Q9QXU9
Gene Alias AI848336; basic protein IA-4; Big LEN; big PEN LEN; Big PEN-LEN; Big SAAS; BigLEN; bLEN; b-LEN; b-PEN-LEN; b-SAAS; granin-like neuroendocrine peptide; IA-4; KEP; Little LEN; little SAAS; lLEN; l-LEN; l-SAAS; N-proSAAS; Pan3; pancreas protein 3; Pcsk1n; PEN; PEN19; PEN-19; PEN20; PEN-20; proprotein convertase 1 inhibitor; Proprotein convertase subtilisin/kexin type 1 inhibitor; proSAAS; pro-SAAS; protein serine alanine alanine serine; SAAS; SAAS CT(1-49); SAAS CT(25-40); SAASCT; SCG8; SgVIII
Gene Symbols PCSK1N
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of rat PCSK1N.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246333
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.