missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCSK9 Monoclonal Antibody (407119)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA524144
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
This antibody detects the mature form of human and mouse Proprotein Convertase 9/PCSK9 in direct ELISAs and Western blots. This antibody does not cross-react with recombinant human (rh) PC-1 or rhPC-7. Reconstitute at 0.5 mg/mL in sterile PBS.
PCSK9 is a proprotein convertase belonging to the proteinase K subfamily of the secretory subtilase family. This protein is synthesized as a soluble zymogen that undergoes autocatalytic intramolecular processing in the endoplasmic reticulum. The protein may function as a proprotein convertase. The protein plays a role in cholesterol homeostasis and may have a role in the differentiation of cortical neurons.
Tekniske data
| PCSK9 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q80W65, Q8NBP7 | |
| PCSK9 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Proprotein Convertase 9/PCSK9 Gln35-Gln694. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Immunoprecipitation, Western Blot | |
| 407119 | |
| Unconjugated | |
| PCSK9 | |
| AI415265; AI747682; convertase subtilisin; convertase subtilisin/kexin type 9 preproprotein; FH3; HCHOLA3; hypercholesterolemia; LDLCQ1; NARC1; NARC-1; neural apoptosis regulated convertase 1; neural apoptosis-regulated convertase 1; OTTHUMP00000009833; PC9; PCSK9; Proprotein convertase 9; proprotein convertase PC9; proprotein convertase subtilisin/kexin type 9; PSEC0052; Subtilisin/kexin-like protease PC9 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 100102, 255738 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion