Learn More
PCSK9 Monoclonal Antibody (OTI8C12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811842S
This item is not returnable.
View return policy
Description
PCSK9 Monoclonal antibody specifically detects PCSK9 in Human samples. It is validated for Western BlotSpecifications
| PCSK9 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8NBP7 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 255738 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI8C12 | |
| Unconjugated | |
| PCSK9 | |
| AI415265; AI747682; convertase subtilisin; convertase subtilisin/kexin type 9 preproprotein; FH3; HCHOLA3; hypercholesterolemia; LDLCQ1; NARC1; NARC-1; neural apoptosis regulated convertase 1; neural apoptosis-regulated convertase 1; OTTHUMP00000009833; PC9; PCSK9; Proprotein convertase 9; proprotein convertase PC9; proprotein convertase subtilisin/kexin type 9; PSEC0052; Subtilisin/kexin-like protease PC9 | |
| Human recombinant protein fragment corresponding to amino acids 153-692 of human PCSK9 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |