missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCTP Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11167-1-AP-20UL
This item is not returnable.
View return policy
Description
Catalyzes the transfer of phosphatidylcholine between membranes. Binds a single lipid molecule.Specifications
| PCTP | |
| Polyclonal | |
| Unconjugated | |
| PCTP | |
| PC TP, PCTP, STARD2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 58488 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.53 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9UKL6 | |
| PCTP | |
| PCTP Fusion Protein Ag1647 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction