missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCYOX1L Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | PCYOX1L |
|---|---|
| Concentration | 300 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18020556
|
Proteintech
30485-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18039375
|
Proteintech
30485-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PCYOX1L antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PCYOX1L. This antibody recognizes human, mouse antigen. The PCYOX1L antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| PCYOX1L | |
| Western Blot, ELISA | |
| Rabbit | |
| 78991 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
| 300 μg/mL | |
| Liquid | |
| Human, Mouse | |
| PCYOX1L, prenylcysteine oxidase 1 like, Prenylcysteine oxidase like | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title