Learn More
Invitrogen™ PCYT1A Recombinant Rabbit Monoclonal Antibody (24GB5015)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557547
 View more versions of this product
Product Code. 30275571
  • € 204.00 / 20µL
Description
PCYT1A Recombinant Monoclonal Antibody for Western Blot, Flow
This gene belongs to the cytidylyltransferase family and is involved in the regulation of phosphatidylcholine biosynthesis. Mutations in this gene are associated with spondylometaphyseal dysplasia with cone-rod dystrophy. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PCYT1A
Recombinant Monoclonal
0.6 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P49585, P49586
PCYT1A
A synthesized peptide derived from human CTA (aa 8-21).
20 μL
Primary
Human, Mouse
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB5015
Unconjugated
PCYT1A
CCT A; CCTA; Cctalpha; CCT-alpha; choline phosphate cytidylyltransferase 1 alpha; choline-phosphate cytidylyltransferase A; CT; CT A; CTA; CTalpha; CTP; CTP phosphocholine cytidylyl transferase; CTP:phosphocholine cytidylyl transferase; CTP:phosphocholine cytidylyltransferase A; CTP:phosphocholine cytidylyltransferase alpha; Ctpct; Cttalpha; PCYT1; PCYT1A; phosphate cytidylyltransferase 1, choline, alpha; phosphate cytidylyltransferase 1, choline, alpha isoform; phosphorylcholine transferase A; SMDCRD
Rabbit
Affinity chromatography
RUO
13026, 5130
-20°C