missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCYT1B Antibody (F39 P7 F11), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-29807CL1
This item is not returnable.
View return policy
Description
PCYT1B Monoclonal antibody specifically detects PCYT1B in Human samples. It is validated for Western Blot
Specifications
| PCYT1B | |
| Monoclonal | |
| CoraFluor 1 | |
| CCT B, CCTB, CCT-betaEC 2.7.7.15, choline-phosphate cytidylyltransferase B, CT B, CTB, CTP:phosphocholine cytidylyltransferase B, EC 2.7.7, phosphate cytidylyltransferase 1, choline, beta, phosphate cytidylyltransferase 1, choline, beta isoform, Phosphorylcholine transferase B | |
| Synthetic Peptide corresponding to the C-Terminus of Human CCT-beta. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| F39 P7 F11 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 9468 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction