missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PCYT1B Polyclonal Antibody
GREENER_CHOICE

Product Code. 15700241
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15700241 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15700241 Supplier Invitrogen™ Supplier No. PA552285

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: PVVTTDAESE TGIPKSLSNE PPSETMEEIE HTCPQPRLTL TAPAPFADET NCQCQAPHEK LTIAQARLGT PADRPVRVYA DGIFDLFHSG HARALMQAKT LFPNSYLLVG V Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 96%.

The protein encoded by this gene belongs to the cytidylyltransferase family. It is involved in the regulation of phosphatidylcholine biosynthesis. Several alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCYT1B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Pcyt1b
Gene Accession No. Q9Y5K3
Gene Alias AW045697; CCT B; CCTB; Cctbeta; CCT-beta; choline phosphate cytidylyltransferase 1 beta; choline-phosphate cytidylyltransferase B; CT B; CTB; CTP:phosphocholine cytidylyltransferase B; CTTbeta; PCYT1B; phosphate cytidylyltransferase 1, choline, beta; phosphate cytidylyltransferase 1, choline, beta isoform; phosphorylcholine transferase B
Gene Symbols Pcyt1b
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PCYT1B. Recombinant protein control fragment (Product #RP-92188).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9468
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.