missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCYT1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13765-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the cytidylyltransferase family. It is involved in the regulation of phosphatidylcholine biosynthesis. Several alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
| PCYT1B | |
| Polyclonal | |
| Unconjugated | |
| PCYT1B | |
| CCT B, CCT beta, CCTB, CT B, CTB, PCYT1B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 236899, 286936, 9468 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q811Q9, Q9QZC4, Q9Y5K3 | |
| Pcyt1b | |
| PCYT1B Fusion Protein Ag4748 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction