missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-1 Antibody (1002313) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8578J
This item is not returnable.
View return policy
Description
PD-1 Monoclonal antibody specifically detects PD-1 in Human, Cynomolgus Monkey samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| PD-1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human, Cynomolgus Monkey | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 1002313 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
| Human embryonic kidney cell, HEK293-derived cynomolgus monkey PD-1, Leu25-Gln167, Accession # NP_001271065.1 | |
| 0.1 mL | |
| Apoptosis, Protein Phosphatase, Stem Cell Markers | |
| 5133 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction