Learn More
PD-1 Antibody (1026519), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB10866MFV610
Product Code. 18501306
  • € 597.00 / 0.10mL
Description
PD-1 Monoclonal antibody specifically detects PD-1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF