missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-1 Antibody (766104) [Alexa Fluor™ 532], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB7738X
This item is not returnable.
View return policy
Description
PD-1 Monoclonal antibody specifically detects PD-1 in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| PD-1 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 766104 | |
| Flow Cytometry, CyTOF-ready | |
| CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse PD-1, Leu25-Gln167, Accession # Q02242 | |
| 0.1 mL | |
| Apoptosis, Protein Phosphatase, Stem Cell Markers | |
| 5133 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction