missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-1 Antibody (Hu121) [Janelia Fluor™ 585], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals FAB11380JF585
This item is not returnable.
View return policy
Description
PD-1 Monoclonal antibody specifically detects PD-1 in Human samples. It is validated for Flow Cytometry
Specifications
| PD-1 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
| Human PD-1 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry | |
| Hu121 | |
| 50mM Sodium Borate | |
| Human | |
| 0.1 mL | |
| Apoptosis, Phospho Specific, Protein Phosphatase, Stem Cell Markers | |
| 5133 | |
| Store at 4°C in the dark. | |
| IgG4 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction