missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-1 Antibody (J43), mFluor Violet 450 SE, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Brand: Novus Biologicals NBP1-43110MFV450
This item is not returnable.
View return policy
Description
PD-1 Monoclonal antibody specifically detects PD-1 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
Specifications
| PD-1 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Armenian Hamster | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| J43 | |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen | |
| CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
| The immunogen for this antibody was PD1. | |
| 0.1 mL | |
| Apoptosis, Phospho Specific, Protein Phosphatase, Stem Cell Markers | |
| 5133 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction