Learn More
Novus Biologicals™ PD-1 Antibody (nofazinlimab) - Humanized, IgG4SP, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28691-1mg
 View more versions of this product
Product Code. 30045796
  • € 920.00 / 1mg
Description
PD-1 Monoclonal antibody specifically detects PD-1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PD-1
Monoclonal
Unconjugated
Q15116
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG4
Flow Cytometry, ELISA, Functional Assay
nofazinlimab
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2
PDCD1/ PD-1/ CD279
1 mg
Apoptosis, Phospho Specific, Protein Phosphatase, Stem Cell Markers
5133