Learn More
PD-1 Antibody (nofazinlimab) [Janelia Fluor™ 669], Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-28691JF669
Product Code. 30173244
  • € 419.00 / 0.10mL
Description
PD-1 Monoclonal antibody specifically detects PD-1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PD-1
Monoclonal
Janelia Fluor 669
CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2
PDCD1/ PD-1/ CD279
0.1 mL
Apoptosis, Phospho Specific, Protein Phosphatase, Stem Cell Markers
5133
Store at 4°C in the dark.
IgG4
Flow Cytometry, ELISA, Functional Assay
nofazinlimab
50mM Sodium Borate
Human
Protein A purified