missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-1 Antibody (SPM597), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47806UV
This item is not returnable.
View return policy
Description
PD-1 Monoclonal antibody specifically detects PD-1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
Specifications
| PD-1 | |
| Monoclonal | |
| DyLight 350 | |
| CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
| Recombinant full-length human PD-1 protein (Uniprot: Q15116) | |
| 0.1 mL | |
| Apoptosis, Stem Cell Markers | |
| 5133 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence | |
| SPM597 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction