missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-1 Mouse anti-Human, Mouse, Rat, DyLight 594, Clone: 7A11B1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-75518DL594
This item is not returnable.
View return policy
Description
PD-1 Monoclonal antibody specifically detects PD-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PD-1 | |
| Monoclonal | |
| DyLight 594 | |
| CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
| A ∼150 amino acid recombinant protein from near the amino terminus of mouse PD-1. aa 22 - 170 | |
| 0.1 mL | |
| Apoptosis, Protein Phosphatase, Stem Cell Markers | |
| 5133 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 7A11B1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction