Learn More
Tocris Bioscience™ PD 166793
GREENER_CHOICE
Product Code. 18035842
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 18035842
Brand: Tocris Bioscience™ 2520/10
Broad spectrum MMP inhibitor
Broad spectrum MMP inhibitor. Displays high affinity for MMP-2, -3 and -13 (IC50 values are 4, 7 and 8 nM respectively) and exhibits > 750-fold selectivity over MMP-1, -7 and -9. Attenuates left ventricular remodelling and dysfunction in rat model of heart failure.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 199850-67-4
Molecular Formula C17H18BrNO4S
Specifications
Quantity 10 mg
Formula Weight 412.3
Purity >99%
Molecular Weight (g/mol) 412.298
Chemical Name or Material PD 166793