missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PD 184352

Description
Selective MEK inhibitor (Ki = 300 nM in vitro). Suppresses FGF-mediated angiogenesis in vivo and decreases VEGF expression. Enhances the therapeutic efficacy of taxol (Cat. No. 1097) in vivo. Orally active.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 478.66 |
| Purity | >98% |
| Molecular Weight (g/mol) | 478.66 |
| Chemical Name or Material | PD 184352 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?