missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-ECGF/Thymidine Phosphorylase Antibody (TYMP/2890R), Alexa Fluor™ 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08402AF350
This item is not returnable.
View return policy
Description
PD-ECGF/Thymidine Phosphorylase Monoclonal antibody specifically detects PD-ECGF/Thymidine Phosphorylase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| PD-ECGF/Thymidine Phosphorylase | |
| Monoclonal | |
| Alexa Fluor 350 | |
| EC 2.4.2, EC 2.4.2.4, ECGF, ECGF1TP, endothelial cell growth factor 1 (platelet-derived), Gliostatin, hPD-ECGF, MEDPS1, MNGIE, MTDPS1, PDECGF, PD-ECGF, Platelet-derived endothelial cell growth factor, TdRPase, thymidine phosphorylase | |
| Recombinant full-length human PD-ECGF/Thymidine Phosphorylase protein (Uniprot: P19971) | |
| 100 μg | |
| Angiogenesis | |
| 1890 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| TYMP/2890R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction