Learn More
PD-L1 Antibody (2096A), PE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals MAB90782PE
Product Code. 18608744
  • € 543.00 / 100µL
Description
PD-L1 Monoclonal antibody specifically detects PD-L1 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PD-L1
Monoclonal
PE
Avelumab, B7-H, B7-H1, B7H1PDCD1L1, CD274 antigenMGC142294, CD274 molecule, PDCD1LG1MGC142296, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1
Mouse myeloma cell line NS0-derived recombinant mouse B7-H1/PD-L1, Met1-Thr239, Accession # Q9EP73
100 μg
Autoimmune Diseases, Cancer, Cellular Markers, Checkpoint signaling, Immune System Diseases, Immunology, Signal Transduction, Tumor Biomarkers, Tumor Suppressors
29126
Store at 4°C in the dark.
IgG
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), CyTOF
2096A